Learn More
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide, 5% w/v in methanol, Thermo Scientific Chemicals
Transesterification reactions
£92.16 - £378.89
Chemical Identifiers
CAS | 68254-41-1 |
---|---|
Molecular Formula | C10H14F3NO |
Molecular Weight (g/mol) | 221.223 |
MDL Number | MFCD00059473 |
InChI Key | BFPOZPZYPNVMHU-UHFFFAOYSA-M |
Synonym | 3-trifluoromethyl phenyltrimethylammonium hydroxide, m-tfptah, trifluoromethylphenyltrimethylammoniumhydroxide, 3-trifluoromethyl phenyltrimethylammonium hydroxide in methanol, n,n,n-trimethyl-3-trifluoromethyl anilinium hydroxide, trimethyl-3-trifluoromethyl phenyl azanium hydroxide, m-trifluoromethylphenyl trimethylammonium hydroxide, trimethyl-3-trifluoromethylphenylammonium hydroxide, n,n,n-trimethyl-3-trifluoromethyl anilinium oxidanide, trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium hydroxide |
PubChem CID | 2760772 |
IUPAC Name | trimethyl-[3-(trifluoromethyl)phenyl]azanium;hydroxide |
SMILES | C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[OH-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11359408
|
Thermo Scientific Alfa Aesar
043284.AA |
5 mL |
£92.16
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349408
|
Thermo Scientific Alfa Aesar
043284.AC |
25 mL |
£378.89
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transesterification reactions.3-(Trifluoromethyl)phenyltrimethylammonium hydroxide is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68254-41-1 | |
221.223 | |
BFPOZPZYPNVMHU-UHFFFAOYSA-M | |
2760772 | |
C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[OH-] |
C10H14F3NO | |
MFCD00059473 | |
3-trifluoromethyl phenyltrimethylammonium hydroxide, m-tfptah, trifluoromethylphenyltrimethylammoniumhydroxide, 3-trifluoromethyl phenyltrimethylammonium hydroxide in methanol, n,n,n-trimethyl-3-trifluoromethyl anilinium hydroxide, trimethyl-3-trifluoromethyl phenyl azanium hydroxide, m-trifluoromethylphenyl trimethylammonium hydroxide, trimethyl-3-trifluoromethylphenylammonium hydroxide, n,n,n-trimethyl-3-trifluoromethyl anilinium oxidanide, trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium hydroxide | |
trimethyl-[3-(trifluoromethyl)phenyl]azanium;hydroxide |
Specifications
68254-41-1 | |
MFCD00059473 | |
3-trifluoromethyl phenyltrimethylammonium hydroxide, m-tfptah, trifluoromethylphenyltrimethylammoniumhydroxide, 3-trifluoromethyl phenyltrimethylammonium hydroxide in methanol, n,n,n-trimethyl-3-trifluoromethyl anilinium hydroxide, trimethyl-3-trifluoromethyl phenyl azanium hydroxide, m-trifluoromethylphenyl trimethylammonium hydroxide, trimethyl-3-trifluoromethylphenylammonium hydroxide, n,n,n-trimethyl-3-trifluoromethyl anilinium oxidanide, trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium hydroxide | |
BFPOZPZYPNVMHU-UHFFFAOYSA-M | |
trimethyl-[3-(trifluoromethyl)phenyl]azanium;hydroxide | |
2760772 | |
5% w/v in methanol | |
3- (Trifluoromethyl)phenyltrimethylammonium hydroxide |
C10H14F3NO | |
UN2924 | |
Miscible with water and alcohols. | |
C[N+](C)(C)C1=CC=CC(=C1)C(F)(F)F.[OH-] | |
221.223 | |
221.22 | |
Liquid |
Safety and Handling
GHS H Statement
H224-H300-H310-H330-H370-H314-H318
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 200-659-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 3-(Trifluoromethyl)phenyltrimethylammonium hydroxide, 5% w/v in methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P311-Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : Methanol
RUO – Research Use Only