missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-3-(Trifluoromethyl)piperidine, 98%, Thermo Scientific™
£123.00 - £359.83
Chemical Identifiers
CAS | 768-31-0 |
---|---|
Molecular Formula | C6H11F3N |
Molecular Weight (g/mol) | 154.16 |
MDL Number | MFCD02183561 |
InChI Key | JOHFJTBDUSVGQB-RXMQYKEDSA-O |
Synonym | 3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl |
PubChem CID | 2760776 |
IUPAC Name | 3-(trifluoromethyl)piperidine |
SMILES | FC(F)(F)[C@@H]1CCC[NH2+]C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15204408
|
Thermo Scientific Acros
454820010 |
1 g |
£123.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15214408
|
Thermo Scientific Acros
454820050 |
5 g | |||||||
Description
Chemical Identifiers
768-31-0 | |
154.16 | |
JOHFJTBDUSVGQB-RXMQYKEDSA-O | |
2760776 | |
FC(F)(F)[C@@H]1CCC[NH2+]C1 |
C6H11F3N | |
MFCD02183561 | |
3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl | |
3-(trifluoromethyl)piperidine |
Specifications
768-31-0 | |
28°C | |
97.5% min. (GC) | |
MFCD02183561 | |
JOHFJTBDUSVGQB-RXMQYKEDSA-O | |
3-(trifluoromethyl)piperidine | |
2760776 | |
98% |
128.0°C to 130.0°C | |
Authentic | |
C6H11F3N | |
3-trifluoromethyl piperidine, dl-3-trifluoromethyl piperidine, piperidine, 3-trifluoromethyl, 3-trifluoromethyl-piperidine, 3-trifluormethyl-piperidine, piperidine,3-trifluoromethyl | |
FC(F)(F)[C@@H]1CCC[NH2+]C1 | |
154.16 | |
153.15 | |
(±)-3-(Trifluoromethyl)piperidine |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (±)-3-(Trifluoromethyl)piperidine, 98%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only