missing translation for 'onlineSavingsMsg'
Learn More

3-Trifluoromethylphenylboronic acid, 97+%, Thermo Scientific Chemicals

Product Code. 10180462
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1423-26-3
Molecular Formula C7H6BF3O2
Molecular Weight (g/mol) 189.93
MDL Number MFCD00151854
InChI Key WOAORAPRPVIATR-UHFFFAOYSA-N
Synonym 3-trifluoromethyl phenylboronic acid, 3-trifluoromethylphenylboronic acid, 3-trifluoromethyl phenyl boronic acid, 3-trifluoromethyl benzeneboronic acid, m-trifluoromethyl phenylboronic acid, 3-trifluoromethylboronic acid, 3-trifluoromethyl phenyl boranediol, 3-boronobenzotrifluoride, 3-trifluoromethyl benzeneboronic acid/anhydride
PubChem CID 2734388
IUPAC Name [3-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC(=C1)C(F)(F)F

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 160°C to 165°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 189.93
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 3-Trifluoromethylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Trifluoromethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'