missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trimethylsilyl)propargyl alcohol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.
Solubility
Not miscible with water.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Protect from heat. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, heat and moisture.
3-(Trimethylsilyl)propargyl alcohol is used as a reagent to synthesize Isbogrel, a Thromboxane A 2 (TXA 2) synthase inhibitor and also a TXA 2 receptor antagonist that has potential to treat asthma.
Solubility
Not miscible with water.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Protect from heat. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, heat and moisture.
Specifications
Boiling Point | 170°C to 172°C |
Quantity | 1 g |
Linear Formula | (CH3)3SiC≡CCH2OH |
UN Number | UN2810 |
Beilstein | 1902505 |
Flash Point | 66°C (150°F) |
Solubility Information | Not miscible with water. |
Formula Weight | 128.25 |
Percent Purity | ≥98% |
Refractive Index | 1.451 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'