Learn More
3-(Trimethylsilyl)propargyl bromide, 97%, Thermo Scientific Chemicals
£39.30 - £72.80
Chemical Identifiers
CAS | 38002-45-8 |
---|---|
Molecular Formula | C6H11BrSi |
Molecular Weight (g/mol) | 191.14 |
MDL Number | MFCD00134460 |
InChI Key | GAPRPFRDVCCCHR-UHFFFAOYSA-N |
Synonym | 3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r |
PubChem CID | 642589 |
IUPAC Name | 3-bromoprop-1-ynyl(trimethyl)silane |
SMILES | C[Si](C)(C)C#CCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10441294
|
Thermo Scientific Acros
399840010 |
1 g |
£39.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10091364
|
Thermo Scientific Acros
399840050 |
5 g |
£72.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
38002-45-8 | |
191.14 | |
GAPRPFRDVCCCHR-UHFFFAOYSA-N | |
642589 | |
C[Si](C)(C)C#CCBr |
C6H11BrSi | |
MFCD00134460 | |
3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r | |
3-bromoprop-1-ynyl(trimethyl)silane |
Specifications
38002-45-8 | |
44°C to 45°C (2.0 mmHg) | |
96% min. (GC) | |
C6H11BrSi | |
MFCD00134460 | |
3-bromo-1-trimethylsilyl-1-propyne, 3-bromoprop-1-yn-1-yl trimethylsilane, 3-bromo-1-propynyl trimethylsilane, 3-trimethylsilyl propargyl bromide, 3-bromoprop-1-ynyl trimethyl silane, silane, 3-bromo-1-propynyl trimethyl, 3-bromoprop-1-yn-1-yl trimethyl silane, acmc-1cthe, ksc496i3r | |
GAPRPFRDVCCCHR-UHFFFAOYSA-N | |
3-bromoprop-1-ynyl(trimethyl)silane | |
642589 | |
97% |
1.3510g/mL | |
62°C | |
Glass Bottle | |
(CH3)3SiC∽CCH2Br | |
1.351 | |
Solubility in water: insoluble | |
C[Si](C)(C)C#CCBr | |
191.14 | |
191.14 | |
3-(Trimethylsilyl)propargyl bromide |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
- 3-(Trimethylsilyl)propargyl bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only