missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals

Product Code. 15480636
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87219-80-5
Molecular Formula C10H20O2Si
Molecular Weight (g/mol) 200.353
MDL Number MFCD08275372
InChI Key MDRSJACTZKYOAL-UHFFFAOYSA-N
Synonym 3,3-diethoxyprop-1-yn-1-yl trimethylsilane, 1-trimethylsilyl-3,3-diethoxy-1-propyne, 3-trimethylsilyl propiolaldehyde diethyl acetal, 3-trimethylsilylpropargyl aldehyde diethyl acetal
PubChem CID 11206442
IUPAC Name 3,3-diethoxyprop-1-ynyl(trimethyl)silane
SMILES CCOC(C#C[Si](C)(C)C)OCC

Specifications

Boiling Point 170°C to 172°C
Quantity 25 g
Linear Formula (CH3)3SiC≡CCH(OCH2CH3)2
UN Number UN1993
Flash Point 40°C (104°F)
Formula Weight 200.35
Percent Purity 97%
Refractive Index 1.431
Chemical Name or Material 3-(Trimethylsilyl)propiolaldehyde diethyl acetal

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'