Learn More
3-(Trimethylsilyl)propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals
POA - £40.10
Chemical Identifiers
CAS | 87219-80-5 |
---|---|
Molecular Formula | C10H20O2Si |
Molecular Weight (g/mol) | 200.353 |
MDL Number | MFCD08275372 |
InChI Key | MDRSJACTZKYOAL-UHFFFAOYSA-N |
Synonym | 3,3-diethoxyprop-1-yn-1-yl trimethylsilane, 1-trimethylsilyl-3,3-diethoxy-1-propyne, 3-trimethylsilyl propiolaldehyde diethyl acetal, 3-trimethylsilylpropargyl aldehyde diethyl acetal |
PubChem CID | 11206442 |
IUPAC Name | 3,3-diethoxyprop-1-ynyl(trimethyl)silane |
SMILES | CCOC(C#C[Si](C)(C)C)OCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15470636
|
Thermo Scientific Alfa Aesar
H53397.06 |
5 g |
£40.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15480636
|
Thermo Scientific Alfa Aesar
H53397.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
87219-80-5 | |
200.353 | |
MDRSJACTZKYOAL-UHFFFAOYSA-N | |
11206442 | |
CCOC(C#C[Si](C)(C)C)OCC |
C10H20O2Si | |
MFCD08275372 | |
3,3-diethoxyprop-1-yn-1-yl trimethylsilane, 1-trimethylsilyl-3,3-diethoxy-1-propyne, 3-trimethylsilyl propiolaldehyde diethyl acetal, 3-trimethylsilylpropargyl aldehyde diethyl acetal | |
3,3-diethoxyprop-1-ynyl(trimethyl)silane |
Specifications
170°C to 172°C | |
C10H20O2Si | |
MFCD08275372 | |
40°C (104°F) | |
MDRSJACTZKYOAL-UHFFFAOYSA-N | |
3,3-diethoxyprop-1-ynyl(trimethyl)silane | |
11206442 | |
97% | |
3-(Trimethylsilyl)propiolaldehyde diethyl acetal |
87219-80-5 | |
(CH3)3SiC≡CCH(OCH2CH3)2 | |
UN1993 | |
3,3-diethoxyprop-1-yn-1-yl trimethylsilane, 1-trimethylsilyl-3,3-diethoxy-1-propyne, 3-trimethylsilyl propiolaldehyde diethyl acetal, 3-trimethylsilylpropargyl aldehyde diethyl acetal | |
CCOC(C#C[Si](C)(C)C)OCC | |
200.353 | |
200.35 | |
1.431 |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only