missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Triphenylphosphoranylidene)tetrahydropyran-2-one, 97%, Thermo Scientific™
£175.00 - £623.50
Chemical Identifiers
CAS | 218282-19-0 |
---|---|
Molecular Formula | C23H21O2P |
Molecular Weight (g/mol) | 360.39 |
MDL Number | MFCD20481847 |
InChI Key | PZGUKMFSICDTKX-UHFFFAOYSA-N |
Synonym | 3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one |
PubChem CID | 10594674 |
IUPAC Name | 3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one |
SMILES | O=C1OCCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11956021
|
Thermo Scientific Acros
440980010 |
1 g |
£175.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11966021
|
Thermo Scientific Acros
440980050 |
5 g | |||||||
Description
Chemical Identifiers
218282-19-0 | |
360.39 | |
PZGUKMFSICDTKX-UHFFFAOYSA-N | |
10594674 | |
O=C1OCCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C23H21O2P | |
MFCD20481847 | |
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one | |
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one |
Specifications
218282-19-0 | |
96% min. (HPLC) | |
C23H21O2P | |
3-triphenyl-??-phosphanylidene oxan-2-one, 3-triphenylphosphoranylidene tetrahydropyran-2-one | |
O=C1OCCCC1=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
360.39 | |
360.39 | |
3-(Triphenylphosphoranylidene)tetrahydropyran-2-one |
228.0°C to 230.0°C | |
Glass bottle | |
MFCD20481847 | |
PZGUKMFSICDTKX-UHFFFAOYSA-N | |
3-(triphenyl-$l^{5}-phosphanylidene)oxan-2-one | |
10594674 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-(3-(Triphenylphosphoranylidene)tetrahydropyran-2-one
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only