Learn More
Masterflex™ 3-Way Large Bore Stopcock
Product Code. 11785069
Click to view available options
Material:
Polycarbonate
Unit Size:
Pack of 10
Product Code. 11785069
Brand: Masterflex™ 3060023
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Perfect for handling higher flow volumes in process while using connections similar to luer systems. Cole-Parmer™ 3-Way Large Bore Stopcock is non-sterile and not approved or intended for medical, clinical, surgical or other patient oriented applications.
  • Enhance fluid flow with a larger bore than standard luer stopcocks
  • Ideal for handling high flow volumes
Specifications
Description 3-way flow
Material Polycarbonate
No. per Pack 10 Pack
Connectors Male Luer Lock, Female Luer