missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ 3-Way Soil Meter

Brannan 3-Way Soil Meter to measure the levels of light, moisture and pH in the soil.

Brand:  Brannan™ 13/400/0

Product Code. 17721328

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Moisture range 1 to 10 (1 to 3: dry/4 to 7: normal/8 to 10: wet)
  • PH range: 3.5 to 8 pH
  • Relative light ranges: 0 to 2000 Lux
  • Robust case
  • 2 x 180 mm metal probes
  • No battery required
Specifications

Specifications

3-Way Soil Meter
Moisture Range 1 to 10
Relative Light Range 0 to 2000 Lux
260 x 60 x 37.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brannan™ 3-Way Soil Meter > Soil Meter; Humidity Range: Moisture Range 1 to 10

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.