Learn More
Masterflex™ 3-way Stopcock
Regulate flow without any leaking with these stopcocks. Cole-Parmer™ Three-Way Stopcocks feature a male luer lock, PC body and an HDPE stem. Handle rotates 180 degrees.
Brand: Masterflex™ 30600-02
Code : A0
Additional Details : Weight : 0.04000kg
Description
Regulate flow without any leaking with these stopcocks. Cole-Parmer™ Three-Way Stopcocks feature a male luer lock, PC body and an HDPE stem. Handle rotates 180 degrees.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applicationsSpecifications
Stopcock | |
Polycarbonate and HDPE | |
Male Luer Lock |
Polycarbonate, HDPE | |
Male Luer Lock |