missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 30.3mm Septum for SCHOTT Screw Caps

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

Brand:  Fisherbrand™ 30 02 0471

622.25 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


Product Code. 11859440

  • £666.00 / Pack of 1000
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • For use with SCHOTT Screw Caps
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Septa
30.3 mm
1000
Beige, Cream
PTFE, Silicone
3.2 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.