Learn More
Tenak™ 30 mm Cryobox Dividers
Dividers for use with 50 mm high cryoboxes.
£34.25 - £47.55
Specifications
Type | Grid Divider |
---|---|
For Use With (Equipment) | 50 mm high cryoboxes |
Height (Metric) | 30 mm |
Product Code | Brand | Tube Array | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Array | Holds | Price | Quantity & Availability | ||||
15210464
|
Tenak™
TE22685 |
8 x 8 cells | Vials with 15.50 mm O.D. |
£45.55
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15210544
|
Tenak™
TE22111 |
3 x 3 cells | Vials with 41 mm O.D. |
£34.25
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15220544
|
Tenak™
TE22112 |
4 x 4 cells | Vials with 31 mm O.D. |
£34.25
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15220624
|
Tenak™
TE22679 |
16 x 16 cells | Vials with 7.50 mm O.D. |
£47.55
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15230544
|
Tenak™
TE22113 |
5 x 5 cells | Vials with 24 mm O.D. |
£34.25
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15230624
|
Tenak™
TE22680 |
14 x 14 cells | Vials with 8.75 mm O.D. |
£47.55
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15240624
|
Tenak™
TE22681 |
13 x 13 cells | Vials with 9.50 mm O.D. |
£47.55
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15250544
|
Tenak™
TE22116 |
13 x 13 cells | Vials with 9 mm O.D. |
£43.45
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15250624
|
Tenak™
TE22682 |
12 x 12 cells | Vials with 10.25 mm O.D. |
£47.55
Pack of 36 |
Estimated Shipment: 30-01-2025
Log in to see stock. |
||||
15260544
|
Tenak™
TE22117 |
14 x 14 cells | Vials with 8 mm O.D. |
£43.45
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15270544
|
Tenak™
TE22118 |
16 x 16 cells | Vials with 7 mm O.D. |
£43.45
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15270624
|
Tenak™
TE22688 |
5 x 5 cells | Vials with 25.25 mm O.D. |
£39.35
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15280624
|
Tenak™
TE22689 |
4 x 4 cells | Vials with 31.75 mm O.D. |
£39.35
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
15290624
|
Tenak™
TE22690 |
3 x 3 cells | Vials with 42.50 mm O.D. |
£39.35
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11541775
|
Tenak™
TE22686 |
7 x 7 cells | Vials with 17.75 mm O.D. |
£43.45
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
11591675
|
Tenak™
TE22115 |
12 x 12 cells | Vials with 10 mm O.D. |
£43.45
Pack of 36 |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
Description
- For use in cardboard cryoboxes
- Holds cryogenic vials
- Grid style divider
Specifications
Grid Divider | |
50 mm high cryoboxes | |
30 mm |