Learn More
TubeWriter 300 - Labeling System
Base Labeling System - Includes 2 Standard Racks
Brand:  TubeWriter TW-300-A
Product Code. 30067427
  • £48900.00 / Each
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Full-featured instrument for printing labels directly on any labware or curved surface
  • Compact footprint for labs with tighter bench space - 20 in. x 18 in. x 18 in. (WxLxH)
  • Designed for the needs of scaling labs
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
Specifications
Specifications
Labeling System
2 Labeling Racks
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TubeWriter 300 - Labeling System > Labeling System