missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Nitrile Class 100 Cleanroom Non-Sterile Gloves (ISO 5)
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
24.46 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Use promo code "21640" to get your promotional price.
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
Description
- Class 100
- Color: White
- Ambidextrous
- Complies with the requirements of the Glove Standard EN420:2003+A1:2009, the European Medical Gloves Standard EN455 (Parts 1, 2 & 3) & EN374-1:2016, EN374-5 (VIRUS)
- Non-sterile
- Powder free, latex free
- Beaded cuff
- Nitrile
- AQL 1.5
Specifications
Certifications/Compliance | ISO 5 |
Certifications/Compliance (Europe) | EN420:2003+A1:2009, the European Medical Gloves Standard EN455 (Parts 1, 2 & 3) & EN374-1:2016 |
Accelerator Free | Yes |
Color | White |
Latex Free | Yes |
Material | Nitrile |
Size | 2X-Large |
Sterility | Non-sterile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'