Learn More
Watson-Marlow Pumps 300 Series Drive with Pumphead
The 300 Series Cased Pump is available as a single or multi-channel benchtop pump with manual, remote, analog, RS232 control and accurate dispensing.
Brand:  Watson-Marlow Pumps 030.3154.3DE
4322.50 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 11341152
  • £4860.00 / Each
Explore more special offers
Description
Description
  • 300 series laboratory pumps are a benchtop pumping solution with manual, remote, analog or RS232 control capabilities.
  • Featuring a chemically resistant ABS top and IP31 protection, 323E/D pumps have a membrane keypad that controls the pump, and a digital LCD to display speed.
  • There are four models with increasing degrees of functionality:
    • 323E/DDu RS232/auto/manual enhanced-control high-flow pumps
    • 323E/DU and 323E/DU analog-auto and enhanced manual-control pumps
    • 323E/DS high-performance manual control pumps
    • 323E/DE manual control pumps
Pumphead options available as standard (others available on request):
  • 313D and 313D2 three-roller pumpheads for continuous tubing
  • 314D and 314D2 four-roller pumpheads for continuous tubing
  • 314MC four-roller five-channel microcassette pumpheads
  • 318MC eight-roller five-channel microcassette pumpheads
  • 501RL and 501RL2 pumpheads for continuous tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Variable Speed
IP31
3 to 400 rpm
323Du/D
100/120/220/240 V
LCD
0.81 to 2000 mL/min.
Yes
313DW