Learn More
TubeWriter 300 - Standard Rack Top Print for 1.5-2.0 mL Microcentrifuge and Screw Cap Tubes
Standard rack for directly printing on the top of 1.5-2.0 mL microcentrifuge and screw cap tubes
Brand: TubeWriter PT-1500MC-300
This item is not returnable.
View return policy
Description
- Standard printing rack designed for the 300
- Prints and labels directly on the top of 1.5 to 2.0 mL microcentrifuge and screw cap tubes
- Rack ensures correct alignment and holds tubes securely
- No time-consuming sticky labels or handwriting required
- Printing can take as little time as 50 tubes in 2 to 3 minutes
- Racks designed for rapid switching between labware types
- Print survives long-term storage at -80°C and liquid nitrogen
- Print withstands the harshest lab conditions
Specifications
Labeling Rack |
Product Content Correction
Product Title
TubeWriter 300 - Standard Rack Top Print for 1.5-2.0 mL Microcentrifuge and Screw Cap Tubes > Standard Rack Top Print for 1.5 to 2.0 mL Microcentrifuge and Screw Cap Tubes
Spot an opportunity for improvement?Share a Content Correction