Learn More
TubeWriter 300 - UV Curable Ink
Replacement UV curable ink
Brand:  TubeWriter TW-UVB300-1
Product Code. 30067437
  • £1060.00 / Each
Estimated Shipment: 04-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ink prints and labels directly on curved plastics and glass surfaces - any labware
  • UV curation ensures printing stands up to harsh laboratory conditions
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Resistant to chemical solvents
  • Disposable cartridges with low replacement costs
  • No time-consuming sticky labels or handwriting required
Specifications
Specifications
Ink Roll Replacement
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TubeWriter 300 - UV Curable Ink > UV Curable Ink