Learn More
RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve, ASTM Certified, Micrometer Pore Sizes
£175.00 - £665.00
Specifications
Diameter (Metric) Outer | 305 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10095081
|
RETSCH
60.159.000125 |
125μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10310962
|
RETSCH
60.159.000106 |
106μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10005091
|
RETSCH
60.159.000150 |
150μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10525262
|
RETSCH
60.159.000212 |
212μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10279211
|
RETSCH
60.159.000250 |
250μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10085011
|
RETSCH
60.159.000025 |
25μm |
£665.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10268821
|
RETSCH
60.159.000355 |
355μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10085021
|
RETSCH
60.159.000032 |
32μm |
£470.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10535262
|
RETSCH
60.159.000300 |
300μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10248831
|
RETSCH
60.159.000038 |
38μm |
£359.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10441952
|
RETSCH
60.159.000500 |
500μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10553522
|
RETSCH
60.159.000425 |
425μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10258831
|
RETSCH
60.159.000045 |
45μm |
£262.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10156171
|
RETSCH
60.159.000053 |
53μm |
£209.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10449841
|
RETSCH
60.159.000600 |
600μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10035051
|
RETSCH
60.159.000063 |
63μm |
£191.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10534302
|
RETSCH
60.159.000075 |
75μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10196061
|
RETSCH
60.159.000850 |
850μm |
£184.00
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
10024761
|
RETSCH
60.159.000710 |
710μm |
£175.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Specifications
305 mm | |
Stainless Steel | |
1Each | |
ASTM E 11 | |
Shaker AS 300 |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
40mm |