Learn More
RETSCH 305 dia. x 40mmH Stainless Steel Test Sieve, ISO Certified, Micrometer Pore Sizes
£175.00 - £665.00
Specifications
Diameter (Metric) Outer | 305 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10648571
|
RETSCH
60.158.000100 |
100μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10544682
|
RETSCH
60.158.000106 |
106μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10595062
|
RETSCH
60.158.000125 |
125μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10486421
|
RETSCH
60.158.000112 |
112μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10496421
|
RETSCH
60.158.000140 |
140μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10370282
|
RETSCH
60.158.000150 |
150μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10467001
|
RETSCH
60.158.000160 |
160μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10762003
|
RETSCH
60.158.000200 |
200μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10259211
|
RETSCH
60.158.000180 |
180μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10136361
|
RETSCH
60.158.000212 |
212μm |
£175.00
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10176501
|
RETSCH
60.158.000224 |
224μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10620642
|
RETSCH
60.158.000025 |
25μm |
£665.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10360572
|
RETSCH
60.158.000280 |
280μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10350572
|
RETSCH
60.158.000250 |
250μm |
£175.00
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
|||||
10759702
|
RETSCH
60.158.000315 |
315μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10468151
|
RETSCH
60.158.000300 |
300μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10759892
|
RETSCH
60.158.000355 |
355μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10771623
|
RETSCH
60.158.000032 |
32μm |
£468.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10514692
|
RETSCH
60.158.000036 |
36μm |
£392.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10238931
|
RETSCH
60.158.000038 |
38μm |
£359.00
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10116651
|
RETSCH
60.158.000400 |
400μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10176641
|
RETSCH
60.158.000040 |
40μm |
£264.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10126651
|
RETSCH
60.158.000425 |
425μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10710473
|
RETSCH
60.158.000450 |
450μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10186641
|
RETSCH
60.158.000045 |
45μm |
£262.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10005151
|
RETSCH
60.158.000500 |
500μm |
£175.00
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
10586212
|
RETSCH
60.158.000050 |
50μm |
£212.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10196641
|
RETSCH
60.158.000053 |
53μm |
£209.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10106031
|
RETSCH
60.158.000560 |
560μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10553722
|
RETSCH
60.158.000056 |
56μm |
£200.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10195971
|
RETSCH
60.158.000071 |
71μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10238831
|
RETSCH
60.158.000630 |
630μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10426431
|
RETSCH
60.158.000710 |
710μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10553912
|
RETSCH
60.158.000063 |
63μm |
£191.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10350812
|
RETSCH
60.158.000600 |
600μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10278971
|
RETSCH
60.158.000800 |
800μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10024791
|
RETSCH
60.158.000075 |
75μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10418731
|
RETSCH
60.158.000080 |
80μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10126121
|
RETSCH
60.158.000090 |
90μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
10436431
|
RETSCH
60.158.000900 |
900μm |
£175.00
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10310912
|
RETSCH
60.158.000850 |
850μm |
£175.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Specifications
305 mm | |
Stainless Steel | |
1Each | |
ISO 3310/1 | |
Shaker AS 300 | |
40mm |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
Certificate indicating Compliance |