missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 305 dia. x 40 mmH Stainless Steel Test Sieve, ISO Certified, Pore Sizes: Under 10 mm

Brand:  RETSCH 60.158.001600

 View more versions of this product

Product Code. 10219171

  • £182.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

1Each
Test Sieve
Stainless Steel
Certificate indicating Compliance
For separation, fractioning, particle size determination
Stainless Steel
1.60 mm
ISO 3310/1
Shaker AS 300
40mm
305 mm
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers