Learn More
Jenway™ 3520 pH/temp meter with glass combination pH electrode
3520 pH/temp meter with glass combination pH electrode, stand and holder, ATC probe, BNC shorting plug, EU power supply offers research grade specifications with a comprehensive range of features and functions, making it suitable for the broadest range of research, general laboratory, QC and GLP based applications.
Brand: Jenway™ 35459-12
990.20 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ideal for quality control and GLP applications
- Up to 3 decimal place resolution
- 1, 2 or 3 point pH calibration
- Automatic or manual buffer selection
- Multiple language options
- Storage of up to 500 results
- RS232 connection to printer or PC or infra red (IrDA) communication
- GLP compliant
- 3 year warranty
Specifications
Glass combination pH electrode, electrode stand and holder, ATC probe, BNC shorting plug, pH 4, 7 and 10 buffer sachets and EU power supply | |
±2 | |
-2 to 20 | |
±1999.9 | |
1, 2 or 3 | |
0.1°C | |
850 g | |
9V AC ± 10, 50/ 60Hz |
±0.5°C | |
±0.003 | |
0.001, 0.01, 0.1 | |
0.1/1mV | |
-10°C to +105°C | |
210 x 250 x 55 mm | |
EU |