Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom, Low Flange, Bar-Coded Microplate
GREENER_CHOICE
Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid
Brand:  Corning™ 3764BC
Product Code. 15565283
  • £1600.71 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Flat bottoms with 112μL total volume
  • Recommended working volumes of 20 to 80μL
  • TC treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
  • Bulk packed 20 per bag
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
16 x 24
Polystyrene
20 to 80 μL
Cell Culture
20/Pk.
With Lid
Black, Clear Bottom
384
Sterile
Square, Flat Bottom (F-well)