missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 384-Well Microtiter Microplates
Excellent for a number of applications.
£309.00
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Product Type | Microplate |
Material | Polystyrene |
No. of Wells | 384 |
Description
Excellent for a number of applications.
These microplates share the same footprint as 96-well versions.
- Square wells
- Design allows for miniaturization of assays and other applications
- Use for high-volume DNA library manipulation and solid-phase binding
Use these microplates for the following application:
- Clear plates—colorimetric applications
- White plates—luminometric/fluorometric applications
- Black plates—fluorometric applications
Specifications
Standard | |
Microplate | |
384 | |
Round Bottom (U-well) | |
50/Cs. |
16 x 24 | |
Polystyrene | |
Non-treated | |
For high throughput screening, PCR, mutagenicity/mutational spectra assays |