missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WebSeal 384-Well Non-Coated Plastic Microplates, U-Shaped Well

For any standard application or assay, you can be sure of excellent solvent resistance and low background noise with these high quality polypropylene 384 well microplates.

£160.00 - £700.00

Specifications

Format Standard
Array 16 x 24
Product Type Microplate
Material Polypropylene
No. of Wells 384
View More Specs

Products 4
Product Code Brand Total Volume (Metric) Well Well Shape No. per Pack Price Quantity & Availability  
Product Code Brand Total Volume (Metric) Well Well Shape No. per Pack Price Quantity & Availability  
15102709
View Documents
Thermo Scientific™
60180-P106
58 μL Round Bottom (U-well) 25 Microplates This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15112709
View Documents
Thermo Scientific™
60180-P136
58 μL Round Bottom (U-well) 100 Microplates This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15152709
View Documents
Thermo Scientific™
60180-P138
120 μL Round Bottom (U-well) 120 Microplates This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15132709
View Documents
Thermo Scientific™
60180-P137
252 μL Round Bottom (U-well) 60 Microplates This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Whatever your standard or routine application or assay, you can be sure of excellent solvent resistance and low background noise with Thermo Scientific™ WebSeal 384-Well Microplates. Made of high-quality, GC-tested polypropylene, these microplates are ideal for:

  • Pharmaceutical and industrial QA/QC
  • High-throughput screening (HTS)
  • Sample collection and storage
  • Combinatorial chemistry
  • Liquid phase assays

Chemically and Thermally Resistant Polypropylene

  • Broad chemical compatibility with alcohols, acetonitrile and other common HPLC solvents
  • Will tolerate temperatures from −80° C to 121°C
  • GC-tested to ensure low extractables

Broad Instrument Compatibility

  • SBS and ANSI standard footprint design

Well Formats to Suit Every Application

  • Shallow well and deep well formats
  • U- and V- bottom wells ensure optimal sample recovery
Specifications

Specifications

Standard
Microplate
384
Non-coated
16 x 24
Polypropylene
Non-sterile
Round Bottom (U-well)
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.