missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 384-Well Polypropylene Plates
GREENER_CHOICE

Product Code. p-156231 Shop All Fisherbrand Products
Click to view available options
Color:
Natural
Red
Unit Size:
Pack of 120
This item is not returnable. View return policy

Product Code. 11957954

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Feature higher well density for economical use of expensive reagents

  • Ideal for storage of compounds and nucleic acid libraries
  • Chemical-resistant
  • Withstand temperatures from -80° to +80°C (-112° to +176°F)
  • Conform to ANSI™-standard footprint for optimal use in automated handling and liquid-dispensing systems

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI
Format Standard
Array 16 x 24
Color Natural
Product Type 384-Well Storage Microplate
Material Polypropylene
No. of Wells 384
Sterility Non-sterile
For Use With (Application) For storage of compounds and nucleic acid libraries
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.