Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
Brand:  Greiner Bio-One 781900
 View more versions of this product
Product Code. 11866091
  • £539.00 / Pack of 40
Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Higher assay sensitivity with reduced background
Service Plan
No
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Black
384
Flat Bottom (F-well)
16 x 24
Polystyrene
Non-sterile