Learn More
Greiner Bio-One 384-well Polystyrene Non-binding Flat Bottom Microplate
Provides an ultra low non-specific biomolecular binding properties. Greiner Bio-One 384 Well Polystyrene Non-binding Flat Bottom Microplate has long-term surface stability without degradation or leaching.
£533.00 - £574.00
Specifications
Format Standard
Array 16 x 24
Material Polystyrene
No. of Wells 384
Sterility Non-sterile
View More Specs
Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
11866091
View Documents Promotion Details
Greiner Bio-One
781900
Black
£533.00
Pack of 40
Currently Unavailable: We have no information when this product will be available
View alternative products
11893891
View Documents Promotion Details
Greiner Bio-One
781904
White
£574.00
Pack of 40
Description
Description
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Higher assay sensitivity with reduced background
Service Plan
No
Specifications
Specifications
Standard
Polystyrene
Non-sterile
16 x 24
384