Learn More
Uvex™ 3B Chem Classic Chemical Protection Coveralls
645.90 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Highly durable material combined with ultrasonically welded and taped seams ensure an effective barrier and maximum safety
- Impervious to liquids
- Low-noise material
- Free of silicone and paint wetting impairment substances
- Material: Polypropylene non woven laminated with Polypropylene film
- Offers protection against a wide range of chemicals
- Middle finger loops prevent sleeves sliding up the arm
- Self-adhesive zipper flap for optimal protection
- Anti-static properties
- Protection against infectious diseases
- Lightweight and durable material
- Skin-friendly textile grip on inside
- Elasticated waistband for a perfect fit
- Secure and convenient closures thanks to elasticated bands on hood, arms and legs
- Comfortable zip puller
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | Large |
Material | Polypropylene |
Color | Yellow |
Certifications/Compliance | EN 14126, EN 14605 - type 3B, EN 14605 - type 4B, ISO 13982-1 - type 5B, EN 1149-5, EN 1073-2 |