Learn More
Uvex™ 3B Chem Light Chemical Protection Coveralls
190.73 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Light and highly flexible material for high wearer comfort
- Liquid-tight
- Low-noise material
- Free from paint wetting impairment substances and silicone
- Ideal for cleaning work
- Middle finger loops for securing the sleeves
- Optimal protection thanks to self-adhesive zipper flap
- Anti-static properties
- Protection against infectious diseases
- Lined with skin-friendly non-woven material
- Secure and comfortable closures thanks to elasticated bands on hood, arms and legs
- Elasticated waistband for optimal fit
- Two-way zip
Specifications
Size | Small |
Material | Polypropylene |
Color | White, Yellow |
Certifications/Compliance | EN 14126, EN 14605 - type 3B, EN 14605 - type 4B, ISO 13982-1 - type 5B, EN 13034 - type 6B, EN 1149-5, EN 1073-2, ISO 27065 C3 |