missing translation for 'onlineSavingsMsg'
Learn More

3BDO, MedChemExpress

Product Code. p-200096183
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16492638

Brand: Medchem Express HYU00434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3BDO is a new mTOR activator which can also inhibit autophagy.

3BDO is a new mTOR activator which can also inhibit autophagy.

Specifications

Chemical Name or Material 3BDO
CAS 890405-51-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H17NO5
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (305.50 mM) ∣Ethanol : 20 mg/mL (61.10 mM; Need ultrasonic)
SMILES O=C1OC(COC2=C([N+]([O-])=O)C=CC=C2)CC1CC3=CC=CC=C3
Molecular Weight (g/mol) 327.33
Formula Weight 327.33
Percent Purity 99.91%
Grade Research
Physical Form Solid-Liquid Mixture
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.