missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film
Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
300 L
560 L
Dimensions:
33 cm x 68.6 cm (13 in. x 27 in.)
45.7 cm x 76.2 cm (18 in. x 30 in.)
55.9 cm x 91.4 cm (22 in. x 36 in.)
70 cm x 122 cm (24 in. x 48 in.)
78.7 cm x 124.5 cm (31 in. x 49 in.)
Unit Size:
Each
Description
The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
| Capacity (Metric) | 560 L |
| Material | Polyethylene |
| Content And Storage | Store at room temperature. |
| Dimensions | 78.7 cm x 124.5 cm (31 in. x 49 in.) |
| Outer Packaging | Individually packed in a header bag |
| Quantity | 560 L, 10 per case |
| Sterilization Method | Irradiated |
| Shipping Condition | Room Temperature |
| Product Line | HyPerforma |
| Port Detail | 0 Ports |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction