missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermo Scientific™ 3D Tank Liner, ASI™ 26 film
Single-use tank liners reduce cleaning steps common to re-usable tanks.
£319.50 - £716.20
Specifications
Material | Polyethylene |
---|---|
Content And Storage | Store at room temperature. |
Outer Packaging | Individually packed in a header bag |
Sterilization Method | Irradiated |
Shipping Condition | Room Temperature |
Product Code | Brand | Capacity (Metric) | Dimensions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions | Price | Quantity & Availability | ||||
17850787
|
Thermo Scientific™
B100363-I |
200 L | 55.9 cm x 91.4 cm (22 in. x 36 in.) | ||||||
17890787
|
Thermo Scientific™
B100364-I |
50 L | 33 cm x 68.6 cm (13 in. x 27 in.) | ||||||
17800797
|
Thermo Scientific™
B100373-I |
100 L | 45.7 cm x 76.2 cm (18 in. x 30 in.) | ||||||
17830797
|
Thermo Scientific™
B101459-I |
300 L | 70 cm x 122 cm (24 in. x 48 in.) | ||||||
17860797
|
Thermo Scientific™
B101460-I |
560 L | 78.7 cm x 124.5 cm (31 in. x 49 in.) | ||||||
Description
The Thermo Scientific™ 3D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Polyethylene | |
Individually packed in a header bag | |
Room Temperature | |
0 Ports | |
No Tubing |
Store at room temperature. | |
Irradiated | |
HyPerforma™ | |
ASI 26 |