missing translation for 'onlineSavingsMsg'
Learn More
3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles Product Code.: 12992315
Estimated Shipment: 18-03-2025
to see stock.
Add to basket

3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles

Product Code. 12992315
3X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy

Product Code. 12992315

Brand: 3M™ 7000089666

Estimated Shipment: 18-03-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Coveralls provide light duty protection against infective agents, certain hazardous dusts, and limited liquid splashes. PE laminate material includes micropores that allow heat and moisture to evaporate from the skin and out through the suit for a lightweight and comfortable fit.

  • Type 5/6 Coverall designed to offer cost effective protection against hazardous dusts and limited liquid splashes
  • Protection against EN 14126 Biological agents
  • Made of a soft lightweight non-woven with microporous laminate for splash protection, strength and breathability
  • Certified protection against Type 5/6 hazardous dusts and limited liquid splashes
  • 2 panel hood design, 2-way zip with storm flap, elasticated apertures reduce the risk of skin exposure
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Material Microporous Polyethylene Laminate
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Closure Type 2-way Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Coating Anti-static
Certifications/Compliance EN 13034 Type 6, EN ISO 13982-1 Type 5, EN 1149-1:2006, ASTM D2859-96, CE
Description White Polypropylene/Polyethylene laminate film suit, Nylon on Polyester Braid zipper, Neoprene Rubber elastic, Polyester thread
Latex Free Yes
Model 4510-3 XL
Size (English) Chest 49 to 52 in.
Size (English) Inseam 35 in.
Size (English) Waist 54 in.
Size (Metric) Chest 124.46 to 132.08 cm
Size (Metric) Inseam 88.9 cm
Size (Metric) Waist 137.16 cm
For Use With (Application) Abatement, Remediation, Agriculture, Cleaning and Maintenance, General Industry, Manufacturing, Non-Hazardous Operations, Painting, Pharmaceutical Cleanroom
Height (English) 6.33 to 6.5 ft.
Height (Metric) 194 to 200 cm
Show More Show Less
  • 3M Safety Solutions - Protective coveralls overview

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Hooded Protective Coveralls 4510 Series, Elastic Wrist and Ankles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.