missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Protective Coveralls 4520, Hooded
Coveralls protect against certain light liquid splashes and hazardous dusts while enhancing movement and comfort.
£193.00 - £206.00
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Polyethylene Film |
Color | White |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12632565
|
3M™
7000034761 |
Medium |
£206.00
Pack of 20 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
12642565
|
3M™
7000034762 |
Large |
£206.00
Pack of 20 |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
12642595
|
3M™
7000034763 |
X-Large |
£206.00
Pack of 20 |
Estimated Shipment: 28-01-2025
Log in to see stock. |
|||||
12652595
|
3M™
7000034764 |
2X-Large |
£201.00
Pack of 20 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
12962285
|
3M™
7000089644 |
4X-Large |
£193.00
Pack of 20 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
11923774
|
3M™
7000089642 |
Small |
£193.00
Pack of 20 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
12952285
|
3M™
7000089643 |
3X-Large |
£201.00
Pack of 20 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
Description
- Designed for use in industrial workplaces as a physical barrier to help prevent skin contact from liquid splashes and dust in certain work environments
- Seamless shoulder and sleeve tops help provide increased comfort and fewer potential entry points for contaminants
- Meet ANSI 101-1996 size guidelines
- Meets EN 340, Type 6 Splash - EN 13034/(EN ISO 17491-4), Type 5 Dust - EN ISO 13982-1, Anti-Static, EN 11492 and Nuclear - EN 1073 - 2
- Microporous PE laminate
- Breathable, SMS fabric back panel helps improve air ventilation and reduce heat build-up
- Reinforcement gusset between the legs to help provide increased durability during bending and squatting
- Elastic waist and ankles, knit cuffs, two-way zipper with sealable storm flap
- Hooded style for use with respiratory equipment when extra protection is required
Specifications
General Purpose Coveralls | |
Microporous Polyethylene Film | |
Single-use | |
Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Non-sterile | |
Anti-static | |
CE, EN 13034, EN-ISO 13982-1, EN 1149-1:2006, EN 1073 | |
No | |
Insulation Installation, Asbestos and Lead Abatement, Coal Dust in Power Plants, Woodworking, Metal Polishing, Light-Duty Building Cleaning, General Powder Handling, General Industrial Clean-up and Maintenance, Machine or Vehicle Maintenance and Food Processing |
Unisex | |
White | |
Elastic | |
Elastic, Knit | |
3-panel | |
Serged | |
Dust, Liquid | |
ANSI 101-1996 | |
3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene | |
4520 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title