missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Protective Overshoe Covers
18.45 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Blue
Description
Reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Made from a microporous laminate material with an anti-static treatment.
- Covers are designed with ties and elastic at the calf to help ensure a secure fit
- Distinctive unisex design
- 3M Catalog# 402
Specifications
Size | Universal |
Material | Polypropylene |
Color | Blue |
Disposable | Single-use |
Closure Type | Elastic Ankle |
Model | 402 |
For Use With (Application) | Assembly, Cleaning, Machine Operations |
Resistance | Dry Particulates, Fluid |
Product Type | Boot Covers |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'