missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Disposable Protective Oversleeve 444
108.00 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
White
Description
Helps protect arms against light liquid splashes and hazardous dusts
- Tapered design and elastic around top and bottom of sleeve for secure and easy fit
- Anti-static treated
- 3M Catalog# 444
Specifications
Size | Universal |
Construction | Microporous laminate |
Material | Microporous Fabric |
Color | White |
Disposable | Single-use |
Cuff Style | Elastic |
Model | 444 |
For Use With (Application) | General maintenance, electronics manufacturing, catering, clean rooms, beauty industries, hygiene, laboratories |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'