missing translation for 'onlineSavingsMsg'
Learn More

3M™ Foam Ear Plugs

Product Code. 12940415
Click to view available options
Model:
1100
1100B
1110
K1110
Corded:
Corded
Uncorded
Unit Size:
100 pairs
200 pairs
500 pairs
Pack of 500
This item is not returnable. View return policy

Product Code. 12940415

Brand: 3M™ 7100100638

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Foam ear plugs are an economical choice for disposable hearing protection. Hypoallergenic and a tapered design help provide a noise-reducing seal in the ear canal.

  • Disposable orange ear plugs
  • Made from hypoallergenic material for maximum protection and comfort
  • Smooth, dirt-resistant surface for hygiene
  • Recommended for high noise situations (up to 100 dBA TWA)

Compliance

3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

ANSI S3.1974 Yes
Cord Color Orange
Cord Material Cloth
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Sanding, Welding
High Visibility Yes
Model 1110
Packaging Type Poly bag
Pairs per Package 500 Pairs/Cs
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 29 dB
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.