Learn More
3M KCl Electrolyte Solution, Hamilton™
Replacement and storage electrolytes for optimal sensor health. Hamilton™ 3M KCl Electrolyte Solution helps ease pH and Redox electrode calibration.
Brand:  Hamilton 238036
 View more versions of this product
Product Code. 10504555
  • £78.55 / Each
Explore more special offers
Description
Description
  • Helps with buffer values changing over time
Specifications
Specifications
3 M
Liquid
Electrolyte
KCL Electrode
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M KCl Electrolyte Solution, Hamilton™ > Capacity: 100mL