missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ SSMMS Lab Coat
Click to view available options
Size:
Medium
Large
Description
- Protect against non hazardous dust and liquid splash
- Knitted cuffs for extra comfort
- Includes internal pocket on the wearers left hand side
- Front zip closing
- Breathable material to help reduce heat build up and promote comfortable wear
- SSMMS material
Specifications
Specifications
Product Type | General Purpose Lab Coat |
Size | Medium |
Gender | Unisex |
Material | SMS |
Color | Cool White |
Closure Type | Zipper |
Cuff Style | Knit |
Disposable | Reusable |
Sterility | Non-sterile |
No. of Pockets | 1 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'