Learn More
3M™ Qualitative Fit-Test Kit Accessories
Product Code. 12993731
Click to view available options
For Use With (Application):
Provide secure hood environment
Quantitative Fit Testing
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter
Product Type:
Collar
Fit Test Adapter
Nebulizer
Respirator Test Hood
Unit Size:
Each
Pack of 2
Pack of 3
Product Code. 12993731
Brand: 3M™ 7000051789
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with 3M™ Qualitative Fit Test Apparatus
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
TRUSTED_SUSTAINABILITY
Specifications
Product Type Collar
Certifications/Compliance OSHA
For Use With (Application) Provide secure hood environment
For Use With (Equipment) Qualitative fit test apparatus FT10 and FT30
Model FT-15