Learn More
3M™ Qualitative Fit-Test Kit Accessories
For use with 3M™ Qualitative Fit Test Apparatus
Brand:  3M™ 7000051788
 View more versions of this product
Product Code. 15749767
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Test kits and accessories designed to help your organization meet OSHA fit testing requirements for respirators
  • The products can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
  • 3M Catalog# 601, FT-13, FT-14, FT-15
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Respirator Test Hood
Used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate filter
FT-14
OSHA
Qualitative Fit Test Apparatus FT10 and FT30
5/Cs.