Learn More
3M™ Sensitivity Solution FT-31, Bitter
The 3M™ Sensitivity Solution FT-31, Bitter can be used to check face-to-respirator seal on any particulate respirator or gas/vapor respirator with a particulate prefilter
Brand: 3M™ 7100004635
3.29 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 7647-14-5 Weight : 0.06030kg
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death
Description
- Bitter taste sensitivity solution for fit testing
- Used with the 3M™ Qualitative Fit Test Apparatus FT-30
- For occupational use only
- Used to test the sensitivity of the respirator wearer to a bitter solution. It is used with the 3M™ Qualitative Fit Test Apparatus FT 30
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product Misuse may result in sickness or death
Specifications
1.9 oz. | |
OSHA 29 CFR 1910.134, Appendix A | |
Bitter Qualitative Fit Test Kit | |
6/Cs. |
55 mL | |
Automotive, Construction, Food and Beverage Manufacturing, Metal Production and Fabrication, Military Maintenance, Repair and Operation, Mining, Oil and Gas, Pharmaceutical, Transportation | |
FT-31 | |
Sensitivity test solution |