Learn More
3M™ Versaflo™ TR-300 / Speedglas™ TR-300 SG PAPR Accessory, Airflow Indicator
The TR-971 air flow indicator may be used to confirm air flow levels of the TR-600 series PAPR. The TR-971 is designed to enable testing at various elevations and temperatures within the operational range of the PAPR system.
Brand: 3M™ 7000103967
21.50 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The TR-971 air flow indicator may be used to confirm air flow levels of the TR-600 series PAPR. The TR-971 is designed to enable testing at various elevations and temperatures within the operational range of the PAPR system. See the User Instructions for details regarding how to use the TR-971.
- Vortemp Air-Heating valve
- ADF not included
- 3M Catalog# TR-970
Specifications
Reusable | |
Ideal to Confirm Air Flow Levels and to Enable Testing at Various Elevations and Temperatures Within The Operational Range of The Powered Air Purifying Respirator System | |
TR-970 |
Airflow Indicator | |
Versaflo TR-300, Speedglas TR-300 SG PAPR System |