Learn More
3M™ Rugged Comfort 6500 Series Quick Latch Half Facepiece Reusable Respirator
Rugged Comfort reusable respirator combines comfort, durability, and stability with an added quick latch mechanism to easily enable the mask to drop down from face without removing head straps.
Brand:  3M™ 7000128239
 View more versions of this product
Product Code. 15757707
  • £34.85 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Resilient silicone faceseal offers extended facepiece life even in high-heat environments
  • Quick latch design eliminates need to remove a hardhat or faceshield when lowering or raising the mask
  • Sleek, low-profile design
  • Special valve cover helps to direct exhaled breath and moisture downward
  • 3M’s proprietary Cool Flow™ Valve helps to make breathing easier by reducing heat and moisture
  • Fits securely and comfortably on the head
  • Compatible with all 3M™ bayonet-style filters and cartridges:
    • 6000 Series Cartridges
    • 2000, 2200, 7000 or 5000 Series Filters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NIOSH 42 CFR Part 84
Bayonet
Half-Mask Respirator
Silicone
Line Compressed Air System SA Series, 3M™ 6000 Series cartridges and 2000, 2200, 7000 or 5000 Series filters
6502QL
0.37 lb.
Quick Latch Mechanism
OSHA 29 CFR Part 1920.134
Gray, Teal
Medium
Facility maintenance, grinding, machine operations, sanding, welding, demolition, assembly, cleaning
Quick Latch
Yolk and cradle
0.16 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Rugged Comfort 6500 Series Quick Latch Half Facepiece Reusable Respirator > Medium