Learn More
3M™ Versaflo™ Air-Flow Adjustment Cover for Premium Head Suspension
Cover is used to adjust the location of the airflow in the hood by attaching to the end of the breathing tube. For use with S-950 Premium Head Suspension.
Brand: 3M™ 7000000157
8.45 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.02000kg
Description
- For use with S-950 Premium Head Suspension
- Attached to end of breathing tube, ring allows user to adjust location of airflow in hood
- 3M Catalog# S-955
Specifications
OSHA | |
Airflow Adjustment Cover | |
S-955 |
Reusable | |
Premium Head Suspension S-950 |