Learn More
3M™ Standard Chemical Sorbents
Absorb virtually any liquid with these chemical sorbents
Brand:  3M™ 7000052037
335.25 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 12332459
  • £509.00 / Pack of 16
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.
  • Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
  • Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
  • Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
  • Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
  • Mini booms surround and contain spills; pillows soak them up
  • 3M Catalog# P110, P200, P208, P300, P500
These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OSHA 29 CFR 1910.132
8 gal.
Polypropylene
For control of spills of certain hazardous liquids
38 x 17.78 cm (15 x 7 in.)
38 cm
17.78 cm
Yellow
30.3 L
Chemical Sorbent Pillow
P300
15 in.
7 in.