Learn More
3M™ Versaflo™ Length-Adjusting Breathing Tube
Features a flexible helix that adjusts to the appropriate length for the individual wearer.
Brand: 3M™ 7100006955
51.90 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : S8
Additional Details : Weight : 0.24120kg
Description
- QRS (Quick Release Swivel) breathing tubes allow fast and easy connection
- Meets higher respiratory performance requirements with 3M™ headtops
- Special flame-resistant sleeve lengthens life of tube and protects it from flying sparks
- Disposable breathing tube cover is available to help cleaning and decontamination operations
- 3M Catalog# BT-30
Specifications
Breathing Tube | |
Assembly, facility maintenance, grinding, machine operation, painting, sanding | |
Versaflo™ TR-300 Series PAPR | |
BT-30 |