Learn More
3M™ Versaflo™ M-Series PAPR Accessory, Ratchet Size Reducing Pad
Adhesive foam pad is for use with any Versaflo™ M-Series product.
Brand: 3M™ 7100064142
75.55 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- When properly attached to the ratchet on the back of the headband, it can help fit smaller wearers and/or provide additional comfort to any wearer
- 3M Catalog# M-956
Specifications
Reusable | |
Comfort Pad | |
Versaflo™ M-Series PAPR System | |
M-956 |
Foam | |
Helps Fit Smaller Wearers and/or Provide Additional Comfort to Any Wearer | |
4-point textile straps, visor |