Learn More
3M™ Versaflo™ TR-300 Series PAPR Accessory, Prefilter
For installation inside the filter cover of a TR-300 Series PAPR
Brand: 3M™ 7000059880
13.40 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00900kg
Description
- Prefilter can extend the life of the HE filter by capturing large particles that would otherwise clog the HE filter
- 3M Catalog# TR-3600
Specifications
Prefilter | |
Versaflo™ tr-300 powered air purifying respirator | |
10/Pk. |
Ideal for use in all applications where hot particles are not present | |
TR-3600 |